5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Akira Shimazu; Tsukasa Miyazaki; Masatoshi Maeda; Kenichi Ikeda. Relationships between the chemical structures and the solubility, diffusivity, and permselectivity of propylene and propane in 6FDA-based polyimides. Journal of Polymer Science Part B: Polymer Physics. 2000,38,(19), 2525-2536.
Masa-Aki Kakimoto; Masaru Yoneyama; Yoshio Imai. Synthesis and properties of new halogen-substituted polyamides from 5-halo-m-phenylenediamines and both aliphatic and aromatic dicarboxylic acids. Journal of Polymer Science Part A: Polymer Chemistry. 2000, 38,(21), 3911-3918.