N-(2-Hydroxyethyl)hexamethyleneimine, 95%, Thermo Scientific Chemicals
N-(2-Hydroxyethyl)hexamethyleneimine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(2-Hydroxyethyl)hexamethyleneimine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L06497.14
also known as L06497-14
Price (USD)/ Each
122.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
122.00
Add to cart
N-(2-Hydroxyethyl)hexamethyleneimine, 95%, Thermo Scientific Chemicals
Catalog numberL06497.14
Price (USD)/ Each
122.00
-
Add to cart
Chemical Identifiers
CAS20603-00-3
IUPAC Name2-(azepan-1-yl)ethan-1-ol
Molecular FormulaC8H17NO
InChI KeyVMRYMOMQCYSPHS-UHFFFAOYSA-N
SMILESOCCN1CCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Refractive Index1.4820-1.4880 @ 20?C
FormLiquid or viscous liquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.

Solubility
Fully miscible in water.

Notes
Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.
RUO – Research Use Only
Michael J. Fazio. Nucleophilic ring opening of 2-oxazolines with amines: a convenient synthesis for unsymmetrically substituted ethylenediamines.J. Org. Chem.1984,49 (5), 4889-4893.