Thermo Scientific Chemicals

Cyclohexanemethylamine, 98%, Thermo Scientific Chemicals

Catalog number: L06501.14
25 g, Each
Thermo Scientific Chemicals

Cyclohexanemethylamine, 98%, Thermo Scientific Chemicals

Catalog number: L06501.14
25 g, Each
Quantity
Catalog number: L06501.14
also known as L06501-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3218-02-8
IUPAC Name
1-cyclohexylmethanamine
Molecular Formula
C7H15N
InChI Key
AVKNGPAMCBSNSO-UHFFFAOYSA-N
SMILES
NCC1CCCCC1
Form
Liquid
Assay (GC)
≥97.5% (UK-sourced material)
Assay from Supplier's CofA
≥97.5% (US-sourced material)
Comment
Material sourced in UK and US
Identification (FTIR)
Conforms (UK-sourced material)

Description

Cyclohexanemethylamine was used in the synthesis of Schiff?s base ligand, 4-methyl-2,6-bis(cyclohexylmethyliminomethyl)-phenol, which forms hexanuclear zinc(II) complexes. It was used during the synthesis of 1-acyl-4 cycloalkyl/arylsemicarbazides and 1-acyl-5-benzyloxy/hydroxyl carbamoylcarbazides from the nonsteroidal anti-inflammatory drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanemethylamine was used in the synthesis of Schiff′s base ligand, 4-methyl-2,6-bis(cyclohexylmethyliminomethyl)-phenol, which forms hexanuclear zinc(II) complexes. It was used during the synthesis of 1-acyl-4 cycloalkyl/arylsemicarbazides and 1-acyl-5-benzyloxy/hydroxyl carbamoylcarbazides from the nonsteroidal anti-inflammatory drugs.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text