Thermo Scientific Chemicals

5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt, 97%, Thermo Scientific Chemicals

Catalog number: L06555.MD
250 mg, Each
Thermo Scientific Chemicals

5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt, 97%, Thermo Scientific Chemicals

Catalog number: L06555.MD
250 mg, Each
Quantity
Catalog number: L06555.MD
also known as L06555-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100900-07-0
IUPAC Name
sodium 5-[(2-aminoethyl)amino]naphthalene-1-sulfonate
Molecular Formula
C12H13N2NaO3S
InChI Key
HGWRACRQRUQQGH-UHFFFAOYSA-M
SMILES
[Na+].NCCNC1=C2C=CC=C(C2=CC=C1)S([O-])(=O)=O
Appearance (Color)
Off-white to yellow
Mass Spec
Conforms
Purity
≥96.0%
Form
Powder or crystals or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text