2-Chloro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals
2-Chloro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L06623.14
also known as L06623-14
Price (USD)/ Each
42.20
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
42.20
Add to cart
2-Chloro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals
Catalog numberL06623.14
Price (USD)/ Each
42.20
-
Add to cart
Chemical Identifiers
CAS777-37-7
IUPAC Name1-chloro-4-nitro-2-(trifluoromethyl)benzene
Molecular FormulaC7H3ClF3NO2
InChI KeyHQROXDLWVGFPDE-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)C(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5065-1.5105 @ 25?C
Appearance (Color)Clear yellow
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only