Thermo Scientific Chemicals

1-Decanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L06642.18
50 g, Each
Thermo Scientific Chemicals

1-Decanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L06642.18
50 g, Each
Quantity
Catalog number: L06642.18
also known as L06642-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61652-81-1
Melting Point (clear melt)
28.0-35.0?C
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White to pale gray
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

1-Decanesulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decanesulfonyl chloride is used as pharmaceutical intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text