Thermo Scientific Chemicals

Ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: L06654.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: L06654.06
5 g, Each
Quantity
Catalog number: L06654.06
also known as L06654-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15400-53-0
IUPAC Name
ethyl 2-amino-6-oxo-1,6-dihydropyrimidine-5-carboxylate
Molecular Formula
C7H9N3O3
InChI Key
HRRHGLKNOJHIGY-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CN=C(N)NC1=O
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (HPLC)
≥94.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text