5-Ethyl-2-furaldehyde, 98%, Thermo Scientific Chemicals
5-Ethyl-2-furaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Ethyl-2-furaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L06659.06
also known as L06659-06
Price (USD)/ Each
147.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
147.00
Add to cart
5-Ethyl-2-furaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberL06659.06
Price (USD)/ Each
147.00
-
Add to cart
Chemical Identifiers
CAS23074-10-4
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1.0%
Appearance (Color)Clear pale yellow to yellow to red to brown
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5205-1.5245 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only