5-Amino-1H-benzotriazole is used as azole building block.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Amino-1H-benzotriazole is used as azole building block.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only
Deniz Sinirlioglu; Ayse Aslan; Ali Ekrem Muftuoglu andAyhan Bozkurt. Synthesis and proton conductivity studies of methacrylate/methacrylamide-based azole functional novel polymer electrolytes.Journal of Applied Polymer Science.2014,131 39915.