Thermo Scientific Chemicals

Carbachol, 98.3%, Thermo Scientific Chemicals

Catalog number: L06674.06
5 g, Each
Thermo Scientific Chemicals

Carbachol, 98.3%, Thermo Scientific Chemicals

Catalog number: L06674.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L06674.06
also known as L06674-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51-83-2
IUPAC Name
2-(trimethylazaniumyl)ethyl carbamate chloride
Molecular Formula
C6H15ClN2O2
InChI Key
AIXAANGOTKPUOY-UHFFFAOYSA-N
SMILES
[Cl-].C[N+](C)(C)CCOC(N)=O
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Water Content (Karl Fischer Titration)
≤1.0%
Form
Crystals or powder or crystalline powder

Description

Carbachol is a cholinergic agonist and is used for the treatment of glaucoma. It is also used as ophthalmic solution (eye drops) during ophthalmic surgery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Carbachol is an ammonium salt and a choline ester that acts on both muscarinic and nicotinic receptors
• Carbachol activates the sympathetic nervous system; it leads to intracellular increases in Ca2+ and activates associated signaling pathways
• Carbachol is not degraded by the cholinesterases
Application
• Carbachol can inhibit apoptotic cell death, making it useful for the study of cell survival and growth in laboratory settings
• Carbachol can be used for laboratory research of any biological pathway that depends on acetylcholine, including sleep cycles and motor control
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text