Thermo Scientific Chemicals

1,6-Hexanedithiol, 97%, Thermo Scientific Chemicals

Catalog number: L06686.06
5 g, Each
Thermo Scientific Chemicals

1,6-Hexanedithiol, 97%, Thermo Scientific Chemicals

Catalog number: L06686.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L06686.06
also known as L06686-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1191-43-1
IUPAC Name
hexane-1,6-dithiol
Molecular Formula
C6H14S2
InChI Key
SRZXCOWFGPICGA-UHFFFAOYSA-N
SMILES
SCCCCCCS
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Refractive Index
1.5105-1.5155 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .

Solubility
Not miscible in water.

Notes
Incompatible materials are Bases, Oxidizing agents, Reducing agents, Alkali metals. Avoid Heat and direct sunlight.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text