Thermo Scientific Chemicals

5-Amino-1H-indazole, 98%, Thermo Scientific Chemicals

Catalog number: L06705.06
5 g, Each
Thermo Scientific Chemicals

5-Amino-1H-indazole, 98%, Thermo Scientific Chemicals

Catalog number: L06705.06
5 g, Each
Quantity
Catalog number: L06705.06
also known as L06705-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19335-11-6
IUPAC Name
1H-indazol-5-amine
Molecular Formula
C7H7N3
InChI Key
XBTOSRUBOXQWBO-UHFFFAOYSA-N
SMILES
NC1=CC=C2NN=CC2=C1
Appearance (Color)
White to red to brown or purple to pink to gray
Form
Crystals or powder or crystalline powder or lumps
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text