Ethyl 2-methylbutyrate, 98%, Thermo Scientific Chemicals
Ethyl 2-methylbutyrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-methylbutyrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number L06743.AP
also known as L06743-AP
Price (USD)/ Each
88.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
88.00
Add to cart
Ethyl 2-methylbutyrate, 98%, Thermo Scientific Chemicals
Catalog numberL06743.AP
Price (USD)/ Each
88.00
-
Add to cart
Chemical Identifiers
CAS7452-79-1
IUPAC Nameethyl 2-methylbutanoate
Molecular FormulaC7H14O2
InChI KeyHCRBXQFHJMCTLF-UHFFFAOYNA-N
SMILESCCOC(=O)C(C)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.3965-1.4005 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only