Thermo Scientific Chemicals

Ethyl vinyl ketone, 97%, stab., Thermo Scientific Chemicals

Catalog number: L06829.06
5 g, Each
Thermo Scientific Chemicals

Ethyl vinyl ketone, 97%, stab., Thermo Scientific Chemicals

Catalog number: L06829.06
5 g, Each
Quantity
Catalog number: L06829.06
also known as L06829-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1629-58-9
IUPAC Name
pent-1-en-3-one
Molecular Formula
C5H8O
InChI Key
JLIDVCMBCGBIEY-UHFFFAOYSA-N
SMILES
CCC(=O)C=C
Form
Liquid
Comment
Stabilized with BHT
Assay from Suppliers CofA
≥96.0%
Appearance (Color)
Clear colorless

Description

Ethyl vinyl ketone is used as reagent for annulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl vinyl ketone is used as reagent for annulation.

Solubility
Insoluble in water; soluble in most organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text