3-Benzyloxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
3-Benzyloxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Benzyloxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L06830.18
also known as L06830-18
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS6346-05-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Free acid (titration)≤1%
Appearance (Color)White to cream to pale yellow
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only