Thermo Scientific Chemicals

2,2,2-Trichloroethyl chloroformate, 97%, Thermo Scientific Chemicals

Catalog number: L06875.14
25 g, Each
Thermo Scientific Chemicals

2,2,2-Trichloroethyl chloroformate, 97%, Thermo Scientific Chemicals

Catalog number: L06875.14
25 g, Each
Quantity
Catalog number: L06875.14
also known as L06875-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17341-93-4
IUPAC Name
2,2,2-trichloroethyl carbonochloridate
Molecular Formula
C3H2Cl4O2
InChI Key
LJCZNYWLQZZIOS-UHFFFAOYSA-N
SMILES
ClC(=O)OCC(Cl)(Cl)Cl
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Refractive Index
1.4680-1.4720 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.

Solubility
Soluble in Ether, Benzene, Chloroform, Ethanol. Insoluble in water.

Notes
Moisture Sensitive. Desiccate at 4°C. Store under inert gas. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text