Thermo Scientific Chemicals

5-Methylindole, 99%, Thermo Scientific Chemicals

Catalog number: L06893.03
1 g, Each
Thermo Scientific Chemicals

5-Methylindole, 99%, Thermo Scientific Chemicals

Catalog number: L06893.03
1 g, Each
Quantity
Catalog number: L06893.03
also known as L06893-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
614-96-0
IUPAC Name
5-methyl-1H-indole
Molecular Formula
C9H9N
InChI Key
YPKBCLZFIYBSHK-UHFFFAOYSA-N
SMILES
CC1=CC=C2NC=CC2=C1
Appearance (Color)
White to cream to pale yellow
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
56.0-62.0?C
Assay (GC)
≥98.5%

Description

5-Methylindole is used as reactant for preparation of pharmaceutically active 2-oxo-1-pyrrolidine analogues, potential anticancer immunomodulators, IL2-inducible T-cell kinase (ITK) inhibitors and CRTh2 antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylindole is used as reactant for preparation of pharmaceutically active 2-oxo-1-pyrrolidine analogues, potential anticancer immunomodulators, IL2-inducible T-cell kinase (ITK) inhibitors and CRTh2 antagonists.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text