Thermo Scientific Chemicals

2-Amino-4-chloro-6-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: L06896.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-4-chloro-6-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: L06896.06
5 g, Each
Quantity
Catalog number: L06896.06
also known as L06896-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5600-21-5
IUPAC Name
4-chloro-6-methylpyrimidin-2-amine
Molecular Formula
C5H6ClN3
InChI Key
NPTGVVKPLWFPPX-UHFFFAOYSA-N
SMILES
CC1=CC(Cl)=NC(N)=N1
Assay (Aqueous acid-base Titration)
>97.5 to <102.5%
Form
Crystalline powder
Assay (HPLC)
>97.5%
Appearance (Color)
White to cream or pale yellow

Description

2-Amino-4-chloro-6-methylpyrimidine is a nitrification inhibitor. It was used to study the influence of chlorine substitution in pyrimidine ring on proton donor ability of amino group in 2-aminopyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-chloro-6-methylpyrimidine is a nitrification inhibitor. It was used to study the influence of chlorine substitution in pyrimidine ring on proton donor ability of amino group in 2-aminopyrimidine.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text