Thermo Scientific Chemicals

1-Methoxy-2-methyl-1-(trimethylsiloxy)propene, 97%, Thermo Scientific Chemicals

Catalog number: L06931.06
5 g, Each
Thermo Scientific Chemicals

1-Methoxy-2-methyl-1-(trimethylsiloxy)propene, 97%, Thermo Scientific Chemicals

Catalog number: L06931.06
5 g, Each
Quantity
Catalog number: L06931.06
also known as L06931-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
31469-15-5
Form
Liquid
Comment
Specification differs for U.S. and non-U.S. material where indicated
Appearance (Color)
Clear colorless
Refractive Index
1.4140-1.4190 @ 20°C (non-U.S. specification)
Assay (GC)
≥96.0%

Description

1-Methoxy-2-methyl-1-(trimethylsiloxy)propene is used in the synthesis of chiral β-lactams by reacting with (S)-alkylidene(1-arylethyl)amines in the presence of titanium tetrachloride. It acts as a catalyst or initiator in the group-transfer polymerization. It is also used as a versatile reagent in conjugate addition4 and aldol reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methoxy-2-methyl-1-(trimethylsiloxy)propene is used in the synthesis of chiral β-lactams by reacting with (S)-alkylidene(1-arylethyl)amines in the presence of titanium tetrachloride. It acts as a catalyst or initiator in the group-transfer polymerization. It is also used as a versatile reagent in conjugate addition4 and aldol reactions.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible materials are Strong oxidizing agents and Strong acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text