Thermo Scientific Chemicals

3,3-Dimethyl-1,2-epoxybutane, 95%, Thermo Scientific Chemicals

Catalog number: L06951.14
25 g, Each
Thermo Scientific Chemicals

3,3-Dimethyl-1,2-epoxybutane, 95%, Thermo Scientific Chemicals

Catalog number: L06951.14
25 g, Each
Quantity
Catalog number: L06951.14
also known as L06951-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2245-30-9
IUPAC Name
2-tert-butyloxirane
Molecular Formula
C6H12O
InChI Key
HEAYDCIZOFDHRM-UHFFFAOYNA-N
SMILES
CC(C)(C)C1CO1
Form
Liquid
Refractive Index
1.3970-1.4030 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥94.0%

Description

3,3-Dimethyl-1,2-epoxybutane is used as pharm and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1,2-epoxybutane is used as pharm and intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text