Thermo Scientific Chemicals

(Chloromethyl)cyclopropane, 97%, Thermo Scientific Chemicals

Catalog number: L06986.06
5 g, Each
Thermo Scientific Chemicals

(Chloromethyl)cyclopropane, 97%, Thermo Scientific Chemicals

Catalog number: L06986.06
5 g, Each
Quantity
Catalog number: L06986.06
also known as L06986-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5911-08-0
Appearance (Color)
Clear colorless to white to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4330-1.4390 @ 20?C

Description

(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4?-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4′-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text