1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals
1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07033.06
also known as L07033-06
Price (USD)/ Each
94.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
94.10
Add to cart
1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals
Catalog numberL07033.06
Price (USD)/ Each
94.10
-
Add to cart
Chemical Identifiers
CAS19600-63-6
IUPAC Name2-(hex-5-en-1-yl)oxirane
Molecular FormulaC8H14O
InChI KeyUKTHULMXFLCNAV-UHFFFAOYNA-N
SMILESC=CCCCCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4335-1.4385 @ 20?C
FormLiquid
1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Maw-Ling Wang; Venugopal Rajendran. Ultrasound assisted phase-transfer catalytic epoxidation of 1,7-octadiene - A kinetic study. Ultrasonics Sonochemistry. 2007, 14,(1), 46-54.
  2. Xuesong Jiang; Hongjie Xu; Jie Yin. Copolymeric dendritic macrophotoinitiators. Polymer. 2005, 46,(24), 11079-11084.