Thermo Scientific Chemicals

4-Bromopyridine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L07052.06
5 g, Each
Thermo Scientific Chemicals

4-Bromopyridine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L07052.06
5 g, Each
Quantity
Catalog number: L07052.06
also known as L07052-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19524-06-2
IUPAC Name
4-bromopyridine hydrochloridyl
Molecular Formula
C5H4BrClN
InChI Key
PSPIMDWZDKGZBO-UHFFFAOYSA-N
SMILES
[Cl].BrC1=CC=NC=C1
Appearance (Color)
White to cream to yellow to red
Water Content (Karl Fischer Titration)
≤0.75%
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms

Description

4-Bromopyridine hydrochloride is used as a useful synthetic intermediate. It is also used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromopyridine hydrochloride is used as a useful synthetic intermediate. It is also used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in DMSO, Methanol and Water.

Notes
Store at -20°C. Hygroscopic. Incompatible materials are moisture, strong bases, strong oxidizing agents. Protect from humidity and air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text