Thermo Scientific Chemicals

3-Methoxypropionitrile, 99%, Thermo Scientific Chemicals

Catalog number: L07055.AE
100 mL, Each
Thermo Scientific Chemicals

3-Methoxypropionitrile, 99%, Thermo Scientific Chemicals

Catalog number: L07055.AE
100 mL, Each
Quantity
Catalog number: L07055.AE
also known as L07055-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-67-8
IUPAC Name
3-methoxypropanenitrile
Molecular Formula
C4H7NO
InChI Key
OOWFYDWAMOKVSF-UHFFFAOYSA-N
SMILES
COCCC#N
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4020-1.4050 @ 20?C
Appearance (Color)
Clear colorless to white to yellow to pale green

Description

3-Methoxypropionitrile is used as a component in high power lithium ion batteries. It is used as pharmaceutical intermediates. It acts as a good solvent for plastic polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxypropionitrile is used as a component in high power lithium ion batteries. It is used as pharmaceutical intermediates. It acts as a good solvent for plastic polymers.

Solubility
Soluble in water. (540 g/L) at 25°C.

Notes
Moisture sensitive. Incompatible materials are Strong oxidizing agents, Strong acids, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text