2-Mercaptothiazole, 97%, Thermo Scientific Chemicals
2-Mercaptothiazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Mercaptothiazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number L07058.14
also known as L07058-14
Price (USD)/ Each
592.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
592.00
Add to cart
2-Mercaptothiazole, 97%, Thermo Scientific Chemicals
Catalog numberL07058.14
Price (USD)/ Each
592.00
-
Add to cart
Chemical Identifiers
CAS82358-09-6
IUPAC Name2,3-dihydro-1,3-thiazole-2-thione
Molecular FormulaC3H3NS2
InChI KeyOCVLSHAVSIYKLI-UHFFFAOYSA-N
SMILESS=C1NC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to green to brown to gray
FormCrystals or powder or crystalline powder or fused solid or flakes
Assay (HPLC)≥96.0%
Melting Point (clear melt)75.0-84.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only