3-Bromo-5-nitrosalicylaldehyde, 98%, Thermo Scientific Chemicals
3-Bromo-5-nitrosalicylaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-5-nitrosalicylaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number L07071.06
also known as L07071-06
Price (USD)/ Each
156.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
156.00
Add to cart
3-Bromo-5-nitrosalicylaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberL07071.06
Price (USD)/ Each
156.00
-
Add to cart
Chemical Identifiers
CAS16789-84-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow to cream
FormPowder or granules
Assay (HPLC)≥97.5%
Melting Point (clear melt)146.5-152.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only