Thermo Scientific Chemicals

Di-n-hexyl ether, 98%, Thermo Scientific Chemicals

Catalog number: L07076.09
10 g, Each
Thermo Scientific Chemicals

Di-n-hexyl ether, 98%, Thermo Scientific Chemicals

Catalog number: L07076.09
10 g, Each
Quantity
Catalog number: L07076.09
also known as L07076-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-58-3
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5% (UK-sourced material)
Refractive Index
1.4180-1.4220 @ 20?C (UK-sourced material)
Comment
Material sourced in UK and US
Form
Liquid

Description

Dihexyl ether has been used as: supported liquid membrane during hollow-fiber liquid-phase microextraction method for determination of nitrophenolic compounds from atmospheric aerosol particles, an extraction solvent to detect avermectins in stream water by hollow-fiber-assisted liquid-phase microextraction technique coupled with LC-MS/MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dihexyl ether has been used as: supported liquid membrane during hollow-fiber liquid-phase microextraction method for determination of nitrophenolic compounds from atmospheric aerosol particles, an extraction solvent to detect avermectins in stream water by hollow-fiber-assisted liquid-phase microextraction technique coupled with LC-MS/MS.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text