2-Biphenylmethanol, 98%, Thermo Scientific Chemicals
2-Biphenylmethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Biphenylmethanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L07094.06
also known as L07094-06
Price (USD)/ Each
93.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
93.20
Add to cart
2-Biphenylmethanol, 98%, Thermo Scientific Chemicals
Catalog numberL07094.06
Price (USD)/ Each
93.20
-
Add to cart
Chemical Identifiers
CAS2928-43-0
IUPAC Name{[1,1'-biphenyl]-2-yl}methanol
Molecular FormulaC13H12O
InChI KeyVKTQADPEPIVMHK-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or granules
Melting Point (clear melt)45.0-51.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only