2-Pyridinesulfonylacetonitrile, 98%, Thermo Scientific Chemicals
2-Pyridinesulfonylacetonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Pyridinesulfonylacetonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L07114.06
also known as L07114-06
Price (USD)/ Each
304.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
304.00
Add to cart
2-Pyridinesulfonylacetonitrile, 98%, Thermo Scientific Chemicals
Catalog numberL07114.06
Price (USD)/ Each
304.00
-
Add to cart
Specifications
Chemical Name or Material2-Pyridinesulfonylacetonitrile
CAS170449-34-0
Health Hazard 1H302+H312+H332
Health Hazard 2GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only