Thermo Scientific Chemicals

(S)-(+)-Carvone, 96%, Thermo Scientific Chemicals

Catalog number: L07130.22
100 g, Each
Thermo Scientific Chemicals

(S)-(+)-Carvone, 96%, Thermo Scientific Chemicals

Catalog number: L07130.22
100 g, Each
Quantity
Catalog number: L07130.22
also known as L07130-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2244-16-8
IUPAC Name
(5S)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one
Molecular Formula
C10H14O
InChI Key
ULDHMXUKGWMISQ-VIFPVBQESA-N
SMILES
CC(=C)[C@H]1CC=C(C)C(=O)C1
Assay (GC)
≥95.0%
Optical Rotation
58.0 ± 2.0? (neat)
Refractive Index
1.4955-1.5030 @ 20?C
Appearance (Color)
Clear colorless to white to yellow
Form
Liquid

Description

Carvones are used in the food and flavor industry. It is also used in air freshening products. Oils containing carvones are used in aromatherapy and alternative medicine. In organic synthesis, it is used as a starting material for the synthesis of terpenoid quassin natural product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carvones are used in the food and flavor industry. It is also used in air freshening products. Oils containing carvones are used in aromatherapy and alternative medicine. In organic synthesis, it is used as a starting material for the synthesis of terpenoid quassin natural product.

Solubility
Miscible with diethyl ether, ethanol and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong reducing agents. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text