4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine is an inhibitor to study the specificity and kinetics of xanthine oxidase. It is also used as an anti-inflammatory agent via inhibition of xanthine oxidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine is an inhibitor to study the specificity and kinetics of xanthine oxidase. It is also used as an anti-inflammatory agent via inhibition of xanthine oxidase.
Solubility
Soluble in DMSO.
Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
Thomas Spector. Oxypurinol as an inhibitor of xanthine oxidase-catalyzed production of superoxide radical.Biochemical Pharmacology.1988,37 349-352.
D W Puett; M B Forman; C U Cates; B H Wilson; K R Hande; G C Friesinger; R Virmani. Oxypurinol limits myocardial stunning but does not reduce infarct size after reperfusion.Circulation.1987,76 678-686..