Allylamine, 98+%, Thermo Scientific Chemicals
Allylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Allylamine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Catalog number L07162.AP
also known as L07162-AP
Price (USD)/ Each
217.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
256.00 
Save 38.35 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
217.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
256.00 
Save 38.35 (15%)
Add to cart
Allylamine, 98+%, Thermo Scientific Chemicals
Catalog numberL07162.AP
Price (USD)/ Each
217.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
256.00 
Save 38.35 (15%)
-
Add to cart
Chemical Identifiers
CAS107-11-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4190-1.4230 @ 20°C (non-U.S. specification)
Assay (GC)≥98.0%
Allylamine is used as monomer in deposition of the allylamine plasma polymer. It was used in generation of a chemical gradient via plasma co-polymerization of octadiene and allylamine polymer on nanoporous alumina. It was used to investigate the surface reactivity of substrates coated with thin layers of plasma polymerized allylamine films. It was used in a direct monocarboxymethylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylamine is used as monomer in deposition of the allylamine plasma polymer. It was used in generation of a chemical gradient via plasma co-polymerization of octadiene and allylamine polymer on nanoporous alumina. It was used to investigate the surface reactivity of substrates coated with thin layers of plasma polymerized allylamine films. It was used in a direct monocarboxymethylation reaction.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.
RUO – Research Use Only

General References:

  1. Andrew Michelmore; Agnieszka Mierczynska; Zihan Poh; Renee V Goreham; Dusan Losic; Robert D Short; Krasimir Vasilev. Versatile gradients of chemistry, bound ligands and nanoparticles on alumina nanopore arrays.Nanotechnology.2011,22 (41), 415601.
  2. G Petranyi; NS Ryder; A Stutz. Allylamine derivatives: new class of synthetic antifungal agents inhibiting fungal squalene epoxidase.Science.1984,224 (4654), 1239-1241..