Thermo Scientific Chemicals

(-)-4-Hydroxy-D-phenylglycine, 98+%

Catalog number: L07190.14
25 g, Each
Thermo Scientific Chemicals

(-)-4-Hydroxy-D-phenylglycine, 98+%

Catalog number: L07190.14
25 g, Each
Quantity
Catalog number: L07190.14
also known as L07190-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22818-40-2
IUPAC Name
(2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
Molecular Formula
C8H9NO3
InChI Key
LJCWONGJFPCTTL-SSDOTTSWSA-N
SMILES
N[C@@H](C(O)=O)C1=CC=C(O)C=C1
Appearance (Color)
White to pale cream
Water Content (Karl Fischer Titration)
≤1% (non-U.S. specification)
Optical Rotation
-157.5 ± 1.5° (C=1 in 1N HCl)
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0% (non-U.S. specification)
Form
Crystals or powder or crystalline powder

Description

(-)-4-Hydroxy-D-phenylglycine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-4-Hydroxy-D-phenylglycine is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water (5 g/L at 20°C) and 1 mol/l HCl.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text