Thermo Scientific Chemicals

1-Octanethiol, 98%, Thermo Scientific Chemicals

Catalog number: L07195.AP
500 mL, Each
Thermo Scientific Chemicals

1-Octanethiol, 98%, Thermo Scientific Chemicals

Catalog number: L07195.AP
500 mL, Each
Quantity
Catalog number: L07195.AP
also known as L07195-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-88-6
IUPAC Name
octane-1-thiol
Molecular Formula
C8H18S
InChI Key
KZCOBXFFBQJQHH-UHFFFAOYSA-N
SMILES
CCCCCCCCS
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Refractive Index
1.4525-1.4570 @ 20°C
Form
Liquid

Description

1-Octanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text