Thermo Scientific Chemicals

N,N-Dimethylphosphoramidodichloridate, 97%, Thermo Scientific Chemicals

Catalog number: L07231.14
25 g, Each
Thermo Scientific Chemicals

N,N-Dimethylphosphoramidodichloridate, 97%, Thermo Scientific Chemicals

Catalog number: L07231.14
25 g, Each
Quantity
Catalog number: L07231.14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
677-43-0
IUPAC Name
(dimethylamino)phosphonoyl dichloride
Molecular Formula
C2H6Cl2NOP
InChI Key
YNHXBEVSSILHPI-UHFFFAOYSA-N
SMILES
CN(C)P(Cl)(Cl)=O
Phosphorus-31 NMR
>96.0%

Description

N,N-Dimethylphosphoramidodichloridate is used in the synthesis of phosphoramidates as well as (RS)-ethyl N,N-dimethylphosphoramidocyanidate (Nerve agent GA) which is used as a chemical weapon. It forms active intermediates, which react with alcohols in situ to give esters. It is also used to convert primary alcohols to alkyl chlorides. Furthe, it serves as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. In addition to this, it reacts with 1,2 diols to prepare cyclic phosphoric amides followed by alkenes after reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylphosphoramidodichloridate is used in the synthesis of phosphoramidates as well as (RS)-ethyl N,N-dimethylphosphoramidocyanidate (Nerve agent GA) which is used as a chemical weapon. It forms active intermediates, which react with alcohols in situ to give esters. It is also used to convert primary alcohols to alkyl chlorides. Furthe, it serves as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. In addition to this, it reacts with 1,2 diols to prepare cyclic phosphoric amides followed by alkenes after reduction.

Solubility
Miscible with dimethoxyethane.

Notes
Moisture and air sensitive. Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text