Thermo Scientific Chemicals

1,3,5-Benzenetricarbonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: L07255.22
100 g, Each
Thermo Scientific Chemicals

1,3,5-Benzenetricarbonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: L07255.22
100 g, Each
Quantity
Catalog number: L07255.22
also known as L07255-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4422-95-1
IUPAC Name
benzene-1,3,5-tricarbonyl trichloride
Molecular Formula
C9H3Cl3O3
InChI Key
UWCPYKQBIPYOLX-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
Form
Crystals or powder or crystalline powder or fused solid or flakes or clear liquid or viscous liquid as melt
Identification (FTIR)
Conforms
Appearance (Color)
Colorless to white to cream to yellow
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Melting Point (clear melt)
33.0-39.0?C

Description

1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Benzenetricarbonyl chloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.

Solubility
Soluble in chloroform(soluble10%, clear to very slightly hazy, colorless to faintly yellow) and toluene. Solubility in water: Reacts.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents. Incompatible with water, bases and oxidizing agents. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text