Thermo Scientific Chemicals

2-tert-Butylphenol, 99%, Thermo Scientific Chemicals

Catalog number: L07294.22
100 g, Each
Thermo Scientific Chemicals

2-tert-Butylphenol, 99%, Thermo Scientific Chemicals

Catalog number: L07294.22
100 g, Each
Quantity
Catalog number: L07294.22
also known as L07294-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-18-6
IUPAC Name
2-tert-butylphenol
Molecular Formula
C10H14O
InChI Key
WJQOZHYUIDYNHM-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=CC=C1O
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to yellow or pale orange
Refractive Index
1.5215-1.5245 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

2-tert-Butylphenol is used pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-tert-Butylphenol is used pharmaceutical intermediates.

Solubility
Soluble in water (0.23 g/100 mL (20°C)).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text