Thermo Scientific Chemicals

2-Deoxy-D-glucose, 98%

Catalog number: L07338.03
1 g, Each
Thermo Scientific Chemicals

2-Deoxy-D-glucose, 98%

Catalog number: L07338.03
1 g, Each
Quantity
Catalog number: L07338.03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
154-17-6
IUPAC Name
(4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
Molecular Formula
C6H12O5
InChI Key
PMMURAAUARKVCB-PHUJZJCSNA-N
SMILES
OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O
Appearance (Color)
White to cream
Identification (FTIR)
Conforms
Form
Powder or crystals or crystalline powder
Assay (HPLC)
≥97.5%
Optical Rotation
+46.5? ± 2.5? (c=0.5 in Water)

Description

2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text