2-Deoxy-D-glucose, 98%
2-Deoxy-D-glucose, 98%
Thermo Scientific Chemicals

2-Deoxy-D-glucose, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number L07338.14
Price (USD)/ Each
756.65
Online exclusive
840.00 
Save 83.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
756.65
Online exclusive
840.00 
Save 83.35 (10%)
Add to cart
2-Deoxy-D-glucose, 98%
Catalog numberL07338.14
Price (USD)/ Each
756.65
Online exclusive
840.00 
Save 83.35 (10%)
-
Add to cart
Chemical Identifiers
CAS154-17-6
IUPAC Name(4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
Molecular FormulaC6H12O5
InChI KeyPMMURAAUARKVCB-PHUJZJCSNA-N
SMILESOC[C@H]1OC(O)C[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Identification (FTIR)Conforms
FormPowder or crystals or crystalline powder
Assay (HPLC)≥97.5%
Optical Rotation+46.5? ± 2.5? (c=0.5 in Water)
View more
2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.
RUO – Research Use Only

General References:

  1. Jiang, X.; Xin, H.; Ren, Q.; Gu, J.; Zhu, L.; Du, F.; Feng, C.; Xie, Y.; Sha, X.; Fang, X. Nanoparticles of 2-deoxy-d-glucose functionalized poly(ethylene glycol)-co-poly(trimethylene carbonate) for dual-targeted drug delivery in glioma treatment. Biomaterials 2014, 35 (1), 518-529.
  2. Xiong, F.; Zhu, Z.; Xiong, C.; Hua, X.; Shan, X.; Zhang, Y.; Gu, N. Preparation, Characterization of 2-Deoxy-D-Glucose Functionalized Dimercaptosuccinic Acid-Coated Maghemite Nanoparticles for Targeting Tumor Cells. Pharm. Res. 2012, 29 (4), 1087-1097.