Thermo Scientific Chemicals

3-Bromobenzanthrone, tech. 85%, Thermo Scientific Chemicals

Catalog number: L07422.14
25 g, Each
Thermo Scientific Chemicals

3-Bromobenzanthrone, tech. 85%, Thermo Scientific Chemicals

Catalog number: L07422.14
25 g, Each
Quantity
Catalog number: L07422.14
also known as L07422-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81-96-9
IUPAC Name
14-bromotetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(17),2,4,6,9,11,13,15-octaen-8-one
Molecular Formula
C17H9BrO
InChI Key
WVECFEIAZAKUNF-UHFFFAOYSA-N
SMILES
BrC1=C2C=CC=C3C(=O)C4=CC=CC=C4C(C=C1)=C23
Appearance (Color)
Pale green to green to yellow to brown
Assay (HPLC)
≥80.0%
Form
Powder
Melting Point (clear melt)
164.5-173.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text