Thermo Scientific Chemicals

2-(Aminomethyl)benzimidazole dihydrochloride hydrate, 98%, Thermo Scientific Chemicals

Catalog number: L07440.14
25 g, Each
Thermo Scientific Chemicals

2-(Aminomethyl)benzimidazole dihydrochloride hydrate, 98%, Thermo Scientific Chemicals

Catalog number: L07440.14
25 g, Each
Quantity
Catalog number: L07440.14
also known as L07440-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5993-91-9
IUPAC Name
1-(1H-1,3-benzodiazol-2-yl)methanamine dihydrochloride
Molecular Formula
C8H11Cl2N3
InChI Key
HAEYZZSSSUIZAN-UHFFFAOYSA-N
SMILES
Cl.Cl.NCC1=NC2=CC=CC=C2N1
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
3.5-11.5% (c. 0.5 to 1.5 waters)
Appearance (Color)
White to cream to pink to blue to gray
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text