Thermo Scientific Chemicals

Methyl linoleate, 95%, Thermo Scientific Chemicals

Catalog number: L07450.06
5 g, Each
Thermo Scientific Chemicals

Methyl linoleate, 95%, Thermo Scientific Chemicals

Catalog number: L07450.06
5 g, Each
Quantity
Catalog number: L07450.06
also known as L07450-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-63-0
IUPAC Name
methyl (9Z,12Z)-octadeca-9,12-dienoate
Molecular Formula
C19H34O2
InChI Key
WTTJVINHCBCLGX-NQLNTKRDSA-N
SMILES
CCCCCC=C/CC=C/CCCCCCCC(=O)OC
Comment
Specification differs for U.S. and non-U.S. material where indicated
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms (non-U.S. specification)
Form
Liquid

Description

The oxidation of methyl linoleate in organic solution and phosphatidylcholine (PC) in liposomal membranes induced by an azo initiator has often been used as a model for lipid peroxidation. Methyl linoleate may be used as a model compound in oxidation/peroxidation assays to evaluate the anti-peroxidation activity of fullerenes, ellagitannins and other natural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The oxidation of methyl linoleate in organic solution and phosphatidylcholine (PC) in liposomal membranes induced by an azo initiator has often been used as a model for lipid peroxidation. Methyl linoleate may be used as a model compound in oxidation/peroxidation assays to evaluate the anti-peroxidation activity of fullerenes, ellagitannins and other natural products.

Solubility
Soluble in DMF, fat solvents, oils.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text