Thermo Scientific Chemicals

2,5-Dichlorobenzyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L07506.06
5 g, Each
Thermo Scientific Chemicals

2,5-Dichlorobenzyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L07506.06
5 g, Each
Quantity
Catalog number: L07506.06
also known as L07506-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2745-49-5
IUPAC Name
1,4-dichloro-2-(chloromethyl)benzene
Molecular Formula
C7H5Cl3
InChI Key
OMZINLIPPVNUOG-UHFFFAOYSA-N
SMILES
ClCC1=CC(Cl)=CC=C1Cl
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Refractive Index
1.5725-1.5775 @ 20°C

Description

2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text