tert-Butylsulfonylacetonitrile, 98%, Thermo Scientific Chemicals
tert-Butylsulfonylacetonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butylsulfonylacetonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L07525.03
also known as L07525-03
Price (USD)/ Each
76.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
76.90
Add to cart
tert-Butylsulfonylacetonitrile, 98%, Thermo Scientific Chemicals
Catalog numberL07525.03
Price (USD)/ Each
76.90
-
Add to cart
Chemical Identifiers
CAS81536-18-7
IUPAC Name2-(2-methylpropane-2-sulfonyl)acetonitrile
Molecular FormulaC6H11NO2S
InChI KeyZRCYCIRRPIYJOU-UHFFFAOYSA-N
SMILESCC(C)(C)S(=O)(=O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)139.5-145.5?C
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only