Thermo Scientific Chemicals

Vinylacetic acid, 96%, Thermo Scientific Chemicals

Catalog number: L07557.22
100 g, Each
Thermo Scientific Chemicals

Vinylacetic acid, 96%, Thermo Scientific Chemicals

Catalog number: L07557.22
100 g, Each
Quantity
Catalog number: L07557.22
also known as L07557-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
625-38-7
IUPAC Name
but-3-enoic acid
Molecular Formula
C4H6O2
InChI Key
PVEOYINWKBTPIZ-UHFFFAOYSA-N
SMILES
OC(=O)CC=C
Assay (GC)
≥95.0%
Refractive Index
1.4210-1.4270 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Form
Liquid

Description

Vinylacetic acid used in the preparation of strained nitroso Diels-Aldrer adducts which undergo a domino ROC metathesis in the presence of Ru-carbene catalysts and olefins. It is used in the synthesis of polyethylene glycolated boltorn coatings. It was used to study the inhibition of the enzyme-catalyzed conversion of D-tyrosyl- phenylalanyl-glycine to D-tyrosyl-phenylaline amide by non-peptide carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinylacetic acid used in the preparation of strained nitroso Diels-Aldrer adducts which undergo a domino ROC metathesis in the presence of Ru-carbene catalysts and olefins. It is used in the synthesis of polyethylene glycolated boltorn coatings. It was used to study the inhibition of the enzyme-catalyzed conversion of D-tyrosyl- phenylalanyl-glycine to D-tyrosyl-phenylaline amide by non-peptide carboxylic acids.

Solubility
Fully miscible.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with oxidizing agents, heat, Bases, Reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text