Thermo Scientific Chemicals

Acetazolamide, 99%, Thermo Scientific Chemicals

Catalog number: L07562.14
25 g, Each
Thermo Scientific Chemicals

Acetazolamide, 99%, Thermo Scientific Chemicals

Catalog number: L07562.14
25 g, Each
Quantity
Catalog number: L07562.14
also known as L07562-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-66-5
Appearance (Color)
White
Assay (HPLC)
≥98.5%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

Acetazolamide acts as a carbonic anhydrase inhibitor which increases cerebral blood flow. It inhibits water permeability of membranes by interacting with aquaporins. It is used for the medical treatment of glaucoma, epileptic seizure, idiopathic intracranial hypertension, altitude sickness, cystinuria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in NH4OH (50 mg/mL), DMSO, Methanol and slightly soluble in Ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text