Thermo Scientific Chemicals

5-Chloro-3-hydroxypyridine, 99%, Thermo Scientific Chemicals

Catalog number: L07568.14
25 g, Each
Thermo Scientific Chemicals

5-Chloro-3-hydroxypyridine, 99%, Thermo Scientific Chemicals

Catalog number: L07568.14
25 g, Each
Quantity
Catalog number: L07568.14
also known as L07568-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74115-12-1
IUPAC Name
5-chloropyridin-3-ol
Molecular Formula
C5H4ClNO
InChI Key
TUIDQYRZDZRHPQ-UHFFFAOYSA-N
SMILES
OC1=CN=CC(Cl)=C1
Melting Point (clear melt)
157.0-163.0?C
Appearance (Color)
Cream to brown
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Crystals or powder or crystalline powder

Description

5-Chloro-3-hydroxypyridine is used in the preparation of 4-(5-chloropyridin-3-yloxy)isoquinoline

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-3-hydroxypyridine is used in the preparation of 4-(5-chloropyridin-3-yloxy)isoquinoline

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text