Thermo Scientific Chemicals

DL-Lysine monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: L07576.14
25 g, Each
Thermo Scientific Chemicals

DL-Lysine monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: L07576.14
25 g, Each
Quantity
Catalog number: L07576.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
885701-25-7
IUPAC Name
2,6-diaminohexanoic acid hydrate
Molecular Formula
C6H16N2O3
InChI Key
HZRUTVAFDWTKGD-UHFFFAOYNA-N
SMILES
O.NCCCCC(N)C(O)=O
Appearance (Color)
White to pale cream to cream to yellow
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
5.8-15.6% (0.5-1.5 waters)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text