Ethyl 4-nitrophenylglyoxylate, 98+%, Thermo Scientific Chemicals
Ethyl 4-nitrophenylglyoxylate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4-nitrophenylglyoxylate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L07579.03
also known as L07579-03
Price (USD)/ Each
76.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
76.00
Add to cart
Ethyl 4-nitrophenylglyoxylate, 98+%, Thermo Scientific Chemicals
Catalog numberL07579.03
Price (USD)/ Each
76.00
-
Add to cart
Chemical Identifiers
CAS70091-75-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to orange or brown
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (GC)≥98.0%
Melting Point38.5-44.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only