Thermo Scientific Chemicals

2-Bromofluorene, 95%, Thermo Scientific Chemicals

Catalog number: L07589.14
25 g, Each
Thermo Scientific Chemicals

2-Bromofluorene, 95%, Thermo Scientific Chemicals

Catalog number: L07589.14
25 g, Each
Quantity
Catalog number: L07589.14
also known as L07589-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1133-80-8
IUPAC Name
2-bromo-9H-fluorene
Molecular Formula
C13H9Br
InChI Key
FXSCJZNMWILAJO-UHFFFAOYSA-N
SMILES
BrC1=CC=C2C(CC3=CC=CC=C23)=C1
Form
Crystals or Crystalline powder
Appearance (Color)
White to pale yellow or cream
Assay (GC)
>94.0%

Description

2-Bromofluorene acts as a nonintrusive end-capping reagent in order to control molecular weights as well as in the generation of well-defined oligomers. It is used to prepare 2-bromo-9-cyclohepta-2,4,6-trienyl-9H-fluorene and 9,9-dihexylflorene. In addition to this, it is used in the preparation of poly(di-n-hexylfluorene)s end capped with 2-bromofluorene, 2-bromo-9,9-di-n-hexylfluorene and 9-bromoanthracene through Ni(0) mediated polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromofluorene acts as a nonintrusive end-capping reagent in order to control molecular weights as well as in the generation of well-defined oligomers. It is used to prepare 2-bromo-9-cyclohepta-2,4,6-trienyl-9H-fluorene and 9,9-dihexylflorene. In addition to this, it is used in the preparation of poly(di-n-hexylfluorene)s end capped with 2-bromofluorene, 2-bromo-9,9-di-n-hexylfluorene and 9-bromoanthracene through Ni(0) mediated polymerization.

Solubility
Soluble in chloroform and ethyl acetate.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text